Learn More
Karl Hecht™ Glass Pasteur Pipettes
Disposable pipettes with a uniform shape for very precise applications in the laboratory.
Brand:  Karl Hecht™ 40567091
 View more versions of this product
Product Code. 10491122
  • 37.00 € / Pack of 250
Explore more special offers
Description
Description
  • Disposable
  • Acc. to ISO 7712
  • Manufactured of soda glass
  • Long tapered tip
  • With constriction in the upper part for cotton plug
  • Capillary tip inside diameter of approx. 1mm, body approx. 90 x 7mm
  • Short form and long form available
  • 250 pieces/box, 1000 pieces/carton
  • With CE marking
  • 100 pieces/tube; 10 tubes/box
Specifications
Specifications
7 mm
Pipetting
Long Tapered
Single-use
Soda Lime Glass
250/Box, 1000/Carton
150 mm