Learn More
Macherey-Nagel™ Glass Plate Adamant
Brand:  Macherey-Nagel™ 821040.200
 View more versions of this product
Product Code. 12814431
  • 319.00 € / Pack of 200
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
5 to 17 μm
200 Pk.
UV254
∼500 m2/g
60 Å
Glass Plate
0.75 mL/g
5 x 10 cm