missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glass Plates for Electrophoresis Systems

For use in standard flat bed electrophoresis units e.g. in the Multiphor II system

Brand:  Cytiva 18-1102-47

Product Code. 10249734

  • 319.00 € / Pack of 2
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Order Info

  • Come in packs of 2
  • Size (L x W): 7.87 x 10.23 in. (20 x 26cm)
  • Thickness: 0.15 in. (4mm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Electrophoresis System
0.15 in.
7.87 x 10.23 in. (20 x 26 cm)
20 cm
26 cm
Glass Plate
0.4 cm
7.87 in.
10.23 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers