Learn More
Hamilton™ Polilyte Lab Electrode
Maintenance free with the accuracy and stability required by laboratory electrodes. Hamilton™ Polilyte Lab Electrodes are easy to use and universally applicable.
Brand: Hamilton™ 238403
This item is not returnable.
View return policy
Description
- Serial number, certificate
- Well suited for measurements in emulsions and suspensions
- Thanks to the Single Pore, clogging of the liquid junction is virtually impossible
Specifications
Polilyte Lab | |
12 mm | |
Glass |
S7 | |
120 mm |