Learn More
Hach Lange™ Glass Radiometer Analytical PHC2005-8 Combination Red-Rod pH Electrode
Brand:  Hach Lange™ E16M500
Product Code. 10502963
  • 505.00 € / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
BNC
pH Electrode
Hach sension benchtop pH meters
Porous Pin
Refillable: Sat. KCl
Radiometer Analytical PHC2005-8 combination pH electrode with Certificate of Conformity and small bottle of internal electrolyte solution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hach Lange™ Glass Radiometer Analytical PHC2005-8 Combination Red-Rod pH Electrode > Connector Type: BNC