Learn More
Poulten Selfe™ Glass Reverse Flow Viscometers
Product Code. 13141093
Click to view available options
Viscosity Range:
0.4 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
Product Code. 13141093
Brand: Poulten Selfe™ 1641/11
Only null left
Add to basket
Add to basket
Suitable for measuring opaque liquids
Opaque 1,600 to 8,000v(mm2s to 1,cst) 500 Poulten Selfe and Lee
Specifications
Product Type Reverse Flow Viscometer
Viscosity Range 1600 to 8000 cSt