Learn More
Poulten Selfe™ Glass Reverse Flow Viscometers
Product Code. 13151083
Click to view available options
Viscosity Range:
0.4 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
Product Code. 13151083
Brand: Poulten Selfe™ 1641/02
Suitable for measuring opaque liquids
Opaque 240 to 1,200v(mm2s-1,cst) 400 Poulten Selfe and Lee
Specifications
Product Type Reverse Flow Viscometer
Viscosity Range 0.8 to 4 cSt