Learn More
Macherey-Nagel™ Glass RP-18 W/UV254 TLC Plate
Glass RP-18 W/UV254 TLC Plate
Brand: Macherey-Nagel™ 811074
698.96 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
HPTLC Glass Plates RP-18 W UV254 C18-modified silica layer
- Base material: glass plates with silical gel
- Mode: Reversed Phase RP-18 W UV254
- Layer Hardness: Standard, Soft
Adsorbent:
- Silica Gel with octadecyl (C18, ODS, RP18) modification
- Phase RP-18 W UV254
- UV Indicator: Yes
- Particle Type: Fully porous particles (FPP)
- Particle Shape: Irregular
- Mean pore size 60 Å
- Specific pore volume 0.75 mL/g,
- Particle size 2-10 μm
- Binder: Yes
Indicators:
This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).
Binder:
Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Recommended Applications:
Aminophenols, Barbiturates, HPTLC, Nucleobases, Phthalates, Plasticizers, Polycyclic aromatic hydrocarbons (PAH), Preservatives, Reversed phase (RP), Steroids, Tetracyclines, Wettable with water
General TLC Glass Plate Features from Macherey-Nagel:
- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- The plate can be easily labelled for clear identification
- Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
- Increased separation efficiency due to an optimized particle size distribution
- A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber
Specifications
Glass RP-18 W | |
60 Å | |
TLC Plate | |
Aminophenols, Barbiturates, Preservatives, Nucleobases, Polycyclic Aromatic Hydrocarbons, Steroids, Tetracyclines, Plasticizers (Phthalates) | |
∼500 m2/g | |
20 x 20 cm |
5 to 17 μm | |
15 Pk. | |
UV254 | |
0.75 mL/g | |
1 mm |