Learn More
Heidolph Instruments Glassware Set - Vertical Condenser (Standard Distillations)
Vertical condenser sets have the smallest space requirement and are good for smaller spaces
Brand:  Heidolph Instruments 513-00300-00
Product Code. 10005711
  • 1335.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints
  • Ventilation cap without ground joints for easy handling and grease-free operation included
  • A venting valve with refill function is optionally available
  • Suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
Specifications
Specifications
230/240 V 50/60 Hz
115/230 VAC
Evaporation Glassware
Borosilicate Glass