Learn More
Hirschmann™ Hematocrit Tubes, Na-heparinized
Excellent for centrifuging blood in the laboratory. Hirschmann™ Hematocrit Tubes are manufactured from disposable capillary glass.
Brand:  Hirschmann™ 9030208
Product Code. 11682299
  • 217.00 € / Pack of 1000
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Na-heparinized
  • For haematocrit
  • CE acc. to IVD 98/79 EC
Specifications
Specifications
ISO 7550, CE, IVD 98/79 EC
9 μL
Micro-hematocrit Capillary
Glass
32 mm
Single-use