Learn More
Glass Syringes
Use these syringes for a variety of applications. Glass syringes feature a metal tip and are fully interchangeable.
Brand:  Samco G812/04
 View more versions of this product
Product Code. 12399219
  • 53.35 € / Each
Edge
Explore more special offers
Description
Description
  • Glass syringe with metal tip
  • Luer fitting
  • Fully interchangeable
  • Available in a variety of sizes
Specifications
Specifications
Glass
Syringe
10 mL