missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Usbeck Laborgeraete™ Glass Tube Cutter
Construced of diecast zinc with nickel plating, this glass tubing cutter is built to last. Usbeck Laborgeraete™ Glass Tube Cutter is designed to cut glass tubes up to 30mm in diameter.
Brand: Usbeck Laborgeraete™ 5220
This item is not returnable.
View return policy
Description
- Durable and effective
- Diecast zinc
- Nickel plating
- Wheel made from hard metal
- Cuts glass tubes up to 30mm in diameter
Specifications
Glass Tube Cutter | |
Cutting glass pipes up to 30mm in diameter | |
1 Ea. | |
68 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction