Learn More
Poulten Selfe™ Glass U-Tube Reverse Flow Viscometer
Suitable viscometer for measuring transparent and opaque liquids. Poulten Selfe™ Glass U-Tube Reverse Flow Viscometer
Brand:  Poulten Selfe™ 1637/09
Product Code. 13121043
  • 282.00 € / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Type BS/IP/RF
  • BS 188, ASTM D445-IP71, ASTM D2170, ISO 3104, ISO
  • Requires long filling pipette
Specifications
Specifications
U-Tube Reverse Flow Viscometers
Yes
Glass
12 - 25 mL
6000 to 30,000 cSt
Kinematic viscosity: 6,000 to 30,000mm2/s (cSt)
9
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products