Learn More
Poulten Selfe™ Glass Ubbelohde
Suitable for transparent liquids
Brand:  Poulten Selfe™ 1634/13
 View more versions of this product
Product Code. 13131073
  • 227.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.
Specifications
Specifications
Ubbelohde Viscometer
Yes
4
2000 to 10,000 cSt
Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products