Learn More
Hirschmann™ Class A Volumetric Flasks with Rim
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Rim are manufactured from durable DURAN™ glass tubing.
29.05 € - 327.00 €
Specifications
Certifications/Compliance | DIN EN ISO 1042 |
---|---|
Material | Borosilicate Glass, Duran Glass |
Graduated | Yes |
Class | A |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10767081
|
Hirschmann™
2800198 |
10 L |
327.00 €
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
10711181
|
Hirschmann™
2800193 |
1000 mL |
48.55 €
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
11768061
|
Hirschmann™
2800180 |
100 mL |
36.50 €
Pack of 2 |
In Stock
Log in to see stock. |
|||||
11708051
|
Hirschmann™
2800160 |
10 mL |
29.05 €
Pack of 2 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
11778061
|
Hirschmann™
2800185 |
200 mL |
48.60 €
Pack of 2 |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
10084720
|
Hirschmann™
2800194 |
2000 mL |
75.75 €
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
11718051
|
Hirschmann™
2800165 |
20 mL |
29.05 €
Pack of 2 |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
11788061
|
Hirschmann™
2800186 |
250 mL |
49.65 €
Pack of 2 |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
11728051
|
Hirschmann™
2800170 |
25 mL |
29.05 €
Pack of 2 |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
11758061
|
Hirschmann™
2800175 |
50 mL |
32.80 €
Pack of 2 |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
11798061
|
Hirschmann™
2800190 |
500 mL |
62.45 €
Pack of 2 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||||
10430041
|
Hirschmann™
2800197 |
5 L |
173.00 €
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
11772412
|
Hirschmann™
2800152 |
5 mL |
29.05 €
Pack of 2 |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
Description
- With blue graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
DIN EN ISO 1042 | |
Borosilicate Glass, Duran Glass | |
Yes | |
A |