Learn More
Hirschmann™ Class A Volumetric Flasks with Hollow Glass Stopper
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Hollow Glass Stopper are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2820253
 View more versions of this product
Product Code. 11790964
  • 46.75 € / Pack of 2
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground, hollow glass stopper
  • With blue stain graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
5 mL
70 mm
10/19 NS Standard Ground
Yes
Glass
Borosilicate Glass, Duran Glass
±0.04 mL
±0.04 mL
A