Learn More
Hirschmann™ Class A USP Volumetric Flasks
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A USP Volumetric Flasks are manufactured from durable DURAN™ glass tubing and are available with a Batch Certificate of Performance, an Individual USP-Certificate of Performance, and a DAkkS-Certificate.
Brand:  Hirschmann™ 282017027
 View more versions of this product
Product Code. 10343073
  • 45.35 € / Pack of 2
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Blue graduation
  • With dated batch identification
  • With NS ground and polyethylene stopper
Specifications
Specifications
USP
25 mL
110 mm
±0.03 mL
A
Polyethylene
Borosilicate Glass, Duran Glass
±0.03 mL
Yes