Learn More
Hirschmann™ Class A Volumetric Flasks with NS Ground
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with NS Ground are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2820390
 View more versions of this product
Product Code. 10498090
  • 70.65 € / Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground, blue graduation
  • Without stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch information
Specifications
Specifications
DIN EN ISO 1042
Borosilicate Glass, Duran Glass
±0.25 mL
±0.25 mL
A
500 mL
300 mm
19/26 NS Standard Ground
Yes