Learn More
Cole-Parmer™ Glass Weighing Micro Funnels
Ensure complete sample transfer to volumetric flasks with these reusable borosilicate glass weighing funnels. Cole-Parmer™ Glass Weighing Micro Funnels are flat on one side so you can set them on a balance.
Brand: Cole-Parmer™ 34700-00
94.21 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Micro funnel is sized to fit microwell plates.Specifications
Borosilicate Glass | |
Sample Transfer to Volumetric Flasks | |
Weighing Funnel |
Product Content Correction
Product Title
Cole-Parmer™ Glass Weighing Micro Funnels > Material: Borosilicate Glass
Spot an opportunity for improvement?Share a Content Correction