Learn More
Macherey-Nagel™ Glass Wool
Brand:  Macherey-Nagel™ 706201
Product Code. 10736502
  • 104.00 € / 50g
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
50g
Glass Wool
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Glass Wool > For Use With: Used for GC liners, packed GC columns etc.