missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glassy Carbon Crucible for Crystal Growth

Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

557.00 € - 1653.23 €

Specifications

Material Glassy Carbon
For Use With (Application) For Crystal Growth
Product Type Specs Glassy Carbon Crucible
View More Specs

Products 5
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Height (Metric) Description Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Height (Metric) Description Price Quantity & Availability  
15493675
View Documents
Thermo Scientific Chemicals
039015.KT
7 mL 14mm 100mm Bottom Angle: 90 deg. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15403685
View Documents
Thermo Scientific Chemicals
039016.KT
25 mL 19mm 140mm Bottom Angle: 60 deg.
915.56 €
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
15413685
View Documents
Thermo Scientific Chemicals
039017.KT
30 mL 24mm 160mm Bottom Angle: 30 deg.
696.47 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
 
15423685
View Documents
Thermo Scientific Chemicals
039018.KT
125 mL 40mm 150mm Bottom Angle: 90 deg. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15433685
View Documents
Thermo Scientific Chemicals
039019.KT
400 mL 57mm 195mm Bottom Angle: 90 deg.
1653.23 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.

Glassy carbon offers:
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
Specifications

Specifications

Glassy Carbon
For Crystal Growth
Glassy Carbon Crucible
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.