Learn More
Thermo Scientific™ Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Probe adaptor and carrying case for use with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Brand:  Thermo Scientific™ 66002-003
 View more versions of this product
Product Code. 10599165
  • 64.70 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Probe adaptor and carrying case for use with the Thermo Scientific™ GLD Pro Gas Leak Detector GLD Pro.
Soft-sided carrying case also for use with GFM Pro Handheld Equipment.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Small Probe Adapter
GLD Pro Gas Leak Detector
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.