Learn More
Plas Labs™ Glove Box 855-ACB Anaerobic Chamber
Manually control your atmosphere when working with oxygen-sensitive materials in this glove box. Plas Labs™ Glove Box 855-ACB Anaerobic Chamber uses a catalyst heater to reduce any trace entering oxygen to water vapor.
Brand:  Plas Labs™ 855-ACB/EXP
Product Code. 10550293
  • 25705.00 € / Each
Explore more special offers
Description
Description
  • Standard 24-month warranty (not applicable for gloves and consumables)
  • Formed one-piece clear acrylic top
  • Self-sealing quick disconnects enable the operator to change the drying train without disturbing the internal atmosphere
  • Matched die-molded thermoset white bottom
  • Adjustable vacuum gauge on transfer chamber
  • Two vacuum pressure pumps, one each for transfer chamber and drying train
  • Four ground key cock valves for purging
  • Drying train includes polycarbonate canisters containing DesiSphere™
  • Inner door on transfer chamber opens automatically when pressures are neutralized between main chamber and transfer chamber
  • All controls are illuminated
  • Transfer chamber is 12in. long and 11in. in diameter (I.D.)
  • Bright Light system (40,000 hour lamp guarantee)
  • Electrical outlet strip; U.L. & C.S.A. approved, hospital grade with four outlets
  • White ambidextrous Hypalon gloves
  • All clamps are adjustable to compensate for wear
Specifications
Specifications
119 cm
107 cm
60 Hz
1157 L
193 cm
97 cm
79 cm
120 V
135 kg
152 cm