missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLS Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3A12A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66265-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes the K-type mitochondrial glutaminase. The encoded protein is an phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. This protein is primarily expressed in the brain and kidney plays an essential role in generating energy for metabolism, synthesizing the brain neurotransmitter glutamate and maintaining acid-base balance in the kidney. Alternate splicing results in multiple transcript variants.Specifications
GLS | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3Z7P3, O94925, P13264 | |
GLS | |
GLS Fusion Protein Ag24227 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3A12A1 | |
Unconjugated | |
GLS | |
AAD20, GAC, GLS, GLS1, glutaminase, K glutaminase, KIAA0838, L glutamine amidohydrolase | |
Mouse | |
Protein A | |
RUO | |
14660, 24398, 2744, 399525 | |
-20°C | |
Liquid |