missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21829-1-AP-150UL
This item is not returnable.
View return policy
Description
21829-1-AP antibody can also detect the 25 kDa degradation protein in SDS-PAGE.
Facilitative glucose transporter. This isoform may be responsible for constitutive or basal glucose uptake. Has a very broad substrate specificity; can transport a wide range of aldoses including both pentoses and hexoses.Specifications
GLUT1 | |
Polyclonal | |
Unconjugated | |
SLC2A1 | |
DYT17, DYT18, GLUT, GLUT 1, GLUT1, HepG2 glucose transporter, PED, SLC2A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20525, 24778, 6513 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11166, P17809 | |
SLC2A1 | |
GLUT1 Fusion Protein Ag16282 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |