Learn More
Glutaric dialdehyde, 25 wt.% solution in water, Thermo Scientific Chemicals
35.88 € - 265.00 €
Specifications
Chemical Name or Material | Glutaric dialdehyde |
---|---|
CAS | 111-30-8, 7732-18-5 |
Density | 1.0600g/mL |
pH | 3.2 to 4.2 |
Molecular Formula | C5H8O2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10366890
|
Thermo Scientific
119980250 |
25 mL | Glass bottle |
35.88 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295340
|
Thermo Scientific Acros
119980010 |
1 L | Glass Bottle |
48.04 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356890
|
Thermo Scientific
119980025 |
2.5 L | Glass bottle |
97.08 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984771
|
Thermo Scientific Acros
119980100 |
10 L | Plastic drum |
265.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glutaric dialdehyde | |
1.0600g/mL | |
C5H8O2 | |
MFCD00007025 | |
01,411 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
16.4mmHg at 20°C |
111-30-8, 7732-18-5 | |
3.2 to 4.2 | |
OHC(CH2)3CHO | |
01, 776 | |
15, 4508 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.12 | |
Solution |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Very t
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-856-5
- Glutaric dialdehyde, 25 wt.% solution in water
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H290-May be corrosive to metals.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only