missing translation for 'onlineSavingsMsg'
Learn More

Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals

Product Code. 10346940
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.32
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point 182.0°C to 192.0°C
Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Quantity 1 g
Beilstein 04,II,931
Merck Index 15,4511
Specific Rotation Condition − 16.50 (20.00°C c=2,H2O)
Specific Rotation − 16.50
Solubility Information Solubility in water: very soluble. Other solubilities: freely soluble in dilute alcohol and dimethyl-, formamide, almost insoluble in ethanol
Formula Weight 307.32
Percent Purity 98%
Grade Analysis
Physical Form Crystalline Powder
Chemical Name or Material Glutathione, For analysis, Reduced
Show More Show Less
Product Identifier
  • Glutathione reduced

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.