Learn More
Glutathione oxidized, MedChemExpress
MedChemExpress Glutathione oxidized (L-Glutathione oxidized) is produced by the oxidation of glutathione which is a major intracellular antioxidant and detoxifying agent.
Brand:  Medchem Express HY-D0844
Encompass_Preferred
Product Code. 16408916
  • 35.10 € / 100mg
Explore more special offers
Description
Description
Glutathione oxidized (L-Glutathione oxidized) is produced by the oxidation of glutathione which is a major intracellular antioxidant and detoxifying agent.
Specifications
Specifications
Glutathione oxidized
White
C20H32N6O12S2
L-Glutathione oxidized GSSG Oxiglutatione
O=C(NCC(O)=O)[C@H](CSSC[C@@H](C(NCC(O)=O)=O)NC(CC[C@H](N)C(O)=O)=O)NC(CC[C@H](N)C(O)=O)=O
612.63
Research
Metabolism-sugar/lipid metabolism
27025-41-8
Research
100 mg
H2O : 250 mg/mL (408.08 mM; Need ultrasonic)
612.63
98.07%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Glutathione oxidized, MedChemExpress