missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Glutathione Sepharose™ 4B Media
Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. Cytiva Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.
324.00 € - 4922.00 €
Specifications
Material | Glutathione Sepharose 4 B |
---|---|
pH Range | 4 to 13 |
Particle Size | 90 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11594935
|
Cytiva
17-0756-01 |
10 mL |
324.00 €
10mL |
In Stock
Log in to see stock. |
|||||
11590794
|
Cytiva
17-0756-05 |
100 mL |
1923.00 €
100mL |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11504945
|
Cytiva
17-0756-04 |
300 mL |
4922.00 €
300mL |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
Description
- Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
- Recommended flowrate (H2O at room temp.): 75cm/hour
- Stable in presence of all commonly used aqueous buffers
- Stable in pH range 4 to 13
Specifications
Glutathione Sepharose 4 B | |
4 to 13 | |
90 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Glutathione Sepharose 4B, 10 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Ethanol