missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glutathione Sepharose™ High Performance Media

For high resolution purification of Glutathione S transferase (GST) tagged proteins

881.00 € - 2836.00 €

Specifications

Binding Capacity >Approx. 10 mg GST-tagged protein/mL resin
Description Glutathione Sepharose High Performance is an affinity resin recommended for purifying glutathione S-transferase (GST)-tagged proteins when high resolution is the priority.
Flow Rate <600 cm/hr.
Matrix Highly crossed-linked agarose, 6%
pH Range 3 to 12
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11555035
View Documents
Cytiva
17-5279-02
100 mL
2836.00 €
100mL
Estimated Shipment: 26-02-2025
to see stock.
Add to basket
 
10032949
View Documents
Cytiva
17-5279-01
25 mL
881.00 €
25mL
In Stock
to see stock.
Add to basket
 
Description

Description

  • One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
  • GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione
  • Mild purification process preserves protein antigenicity and function
  • Glutathione ligand is coupled to 6% agarose matrix

Specifications

Specifications

>Approx. 10 mg GST-tagged protein/mL resin
<600 cm/hr.
3 to 12
4°C to 30°C, 20% Ethanol
Chromatography Media
Glutathione Sepharose High Performance is an affinity resin recommended for purifying glutathione S-transferase (GST)-tagged proteins when high resolution is the priority.
Highly crossed-linked agarose, 6%
6% Highly Cross-linked Agarose
34 μm
Glutathione
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.