Learn More
MP Biomedicals™ Gly-Phe-7-Amino-4-Trifluoromethylcoumarin
Gly-Phe-7-Amino-4-Trifluoromethylcoumarin, White powder
Brand: MP Biomedicals™ 03AFC03325
This item is not returnable.
View return policy
Description
- Gly-Phe-AFC is a fluorogenic, cell-permeant peptide substrate, used in Cytotoxicity testing.
- In vitro cytotoxicity testing has become an integral aspect of drug discovery because it is a convenient, costeffective, and predictive means of characterizing the toxic potential of new chemical entities.
- It is also used as substrate for measuring Dipeptidyl peptidase1 (DPP1) activity.
Specifications
White | |
Cell Migration Analysis; Cell Signaling Analysis | |
433.38 ; 574 (Including TFA salt) | |
Keep cool and dry |
Powder | |
25mg | |
Proteins, Enzymes & Peptides |