missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 98+%, Extra Pure, SLR, Fisher Chemical™

44.05 € - 585.00 €

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10675872
View Documents
Fisher Chemical
G/0600/08
500 mL HDPE plastic bottle
44.05 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10336040
View Documents
Fisher Chemical
G/0600/17
2.5 L HDPE plastic bottle
171.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10685872
View Documents
Fisher Chemical
G/0600/25
25 L Plastic container
585.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glycerol, 98+%, Extra Pure, SLR

Specifications

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO

Specifications

56-81-5
5
C3H8O3
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
OCC(O)CO
92.09
3.17
92.09g/mol
Very Viscous Liquid
18°C
290°C
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
CHEBI:17754
0.003mbar 50
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.