missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ≥99.5%, Molecular biology reagent grade, MP Biomedicals™

Product Code. 11486441
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Glycerol is a simple polyol compound which is a viscous liquid that is widely used in pharmaceutical formulations. It has three hydroxyl groups that are responsible for its solubility in water and its hygroscopic nature

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.2613g/cm3
Assay Percent Range ≥99.5%
Quantity 500 mL
Formula Weight 92.09
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material Glycerol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.