missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™

Product Code. 10579570
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container

C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
pH 5
Boiling Point 290°C
Packaging HDPE plastic bottle
Quantity 500 mL
Vapor Density 3.17
Formula Weight 92.09g/mol
Vapor Pressure 0.003mbar 50
Physical Form Very Viscous Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'