missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10562524

Glycerol, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10562524
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10562524

Brand: Thermo Scientific Acros 158922500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Decomposition Information >290°C
Melting Point 18.0°C
Density 1.2610g/mL
Boiling Point 290.0°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.4716
Quantity 250 mL
Beilstein 01, 502
Merck Index 15, 4520
Specific Gravity 1.261
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Viscosity 1069 mPa.s (20°C)
Formula Weight 92.09
Vapor Pressure 0.003mbar at 50°C
Percent Purity 99+%
Grade Extra Pure
Physical Form Viscous Liquid
Chemical Name or Material Glycerol, Extra Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2497163 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158922500,158920100,158920010,158920250,158920025
Lot Number2420389 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158922500,158920100,158920010,158920250,158920025
Lot Number2412564 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158922500,158920100,158920010,158920250,158920025
Lot Number2494632 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158922500,158920100,158920010,158920250,158920025
Lot Number2495927 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158922500,158920100,158920010,158920250,158920025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.