Learn More
Glycerol formal, 99+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 259772500
136.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Glycerol formal | |
4740-78-7, 5464-28-8 | |
4.0 to 6.5 (10% soln.) | |
97°C | |
99% min. Sum of alpha-and beta-isomers (GC) | |
C4H8O3 | |
MFCD00003218 | |
14, 4485 | |
(50% in water) Clear | |
16 mPa.s (20°C) | |
99+% |
99+% | |
1.2100g/mL | |
193.0°C to 195.0°C | |
Authentic | |
Glass bottle | |
1.4500 to 1.4520 | |
250 mL | |
1.21 | |
104.11 | |
104.11 | |
Liquid |
Safety and Handling
- Glycerol formal, mixture of approx. 55% 5-Hydroxy-1,3-dioxane and approx. 45% 4-Hydroxymethyl-1,3-dioxolane
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H361-Suspected of damaging fertility or the unborn child.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only