missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents

54.85 € - 74.00 €

Specifications

Chemical Name or Material Glycerol Gel-Loading Dye, 5X
Name Note Contains 30% Glycerol
CAS 56-81-5, 7732-18-5
Absorbance 0.25 to 0.40 AU, 0.25 to 0.45 AU at 252nm, 0.30 to 0.45 AU
Color Colorless
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10205263
View Documents
Fisher BioReagents
BP645-1
1 mL Poly Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10400084
View Documents
Fisher BioReagents
BP645-5
5 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This dye is added to DNA and RNA samples prior to electrophoresis on agarose gels.
Specifications

Specifications

Glycerol Gel-Loading Dye, 5X
56-81-5, 7732-18-5
Colorless
Glycerol (30%), Proprietary Components (<6%), and Water
Contains 30% Glycerol
0.25 to 0.40 AU, 0.25 to 0.45 AU at 252nm, 0.30 to 0.45 AU
Molecular Biology
Liquid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.