missing translation for 'onlineSavingsMsg'
Learn More

Glycerol (Molecular Biology), Fisher BioReagents™

Product Code. 12144481
Click to view available options
Quantity:
1 L
4 L
Packaging:
HDPE Bottle

For low-temperature storage of enzymes and bacterial cultures

Glycerol is used for low-temperature storage of enzymes and bacterial cultures, purification of proteins, and as a component in electrophoresis loading buffers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Decomposition Information >290°C
Melting Point 18°C
Density 1.261g/cm³
pH 5
Boiling Point 290°C
Assay Percent Range ≥99.5 %
Packaging HDPE Bottle
Linear Formula HOCH2CH(OH)CH2OH
Quantity 4 L
Merck Index 15, 4520
Formula Weight 92.09
Vapor Pressure 0.003mbar at 50°C
Percent Purity ≥99.5%
Grade DNase-, RNase- and Protease-Free
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.