missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, puriss. p.a., ACS reagent, Honeywell

Product Code. 15665690
Click to view available options
Quantity:
250 mL
1 L
2.5 L

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.25g/mL
Flash Point 160°C (320°F)
Ignition Point 698°F
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1 mmHg (20°C)
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Glycerol
Show More Show Less
Product Identifier
  • Glycerol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'