missing translation for 'onlineSavingsMsg'
Learn More

Glycerol tributyrate, 97%, Thermo Scientific Chemicals

Product Code. 11433784
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Glycerol tributyrate, 98%, C15H26O6, CAS Number-60-01-5, tri-n-butyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, glycerol tributyrate, glycerol tributanoate, glyceroltributyrin, tributyrin, butyrin, butyryl triglyceride, tributin, 1000g, 1714746, 307 deg.C to 308 deg.C, CHEBI:35020

Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.

Solubility
Not miscible in water.

Notes
Incompatible with strong oxidizing agents, Strong acids, Strong bases. Use with adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-01-5
Molecular Formula C15H26O6
Molecular Weight (g/mol) 302.367
MDL Number MFCD00009392
InChI Key UYXTWWCETRIEDR-UHFFFAOYSA-N
Synonym tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
PubChem CID 6050
ChEBI CHEBI:35020
IUPAC Name 2,3-di(butanoyloxy)propyl butanoate
SMILES CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

Specifications

Melting Point -75°C
Density 1.032
Boiling Point 307°C to 308°C
Flash Point 174°C (345°F)
Refractive Index 1.4358
Quantity 50 g
Beilstein 1714746
Merck Index 14,9620
Solubility Information Not miscible in water.
Formula Weight 302.37
Percent Purity 97%
Chemical Name or Material Glycerol tributyrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'