missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Glycine, 99+% Product Code.: 11327908

Thermo Scientific Chemicals Glycine, 99+%

Product Code. 11327908
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11327908

Brand: Thermo Scientific Chemicals 036435.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed to protect the kidneys from the harmful side effects of certain drugs used after organ transplantation as well as the liver from the harmful effects of alcohol. The solution of glycine plays an important role for deposition of alloys and metals. It is likewise useful for patients with acute alcohol hallucinosis. It is applied for treating schizophrenia, stroke, benign prostatic hyperplasia (BPH), and some rare inherited metabolic disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point ∽245°C (decomposition)
Assay Percent Range 99.5+%
Beilstein 635782
Merck Index 14,4491
Solubility Information Soluble in pyridine,water. Sparingly soluble in ethanol. Insoluble in ether.
Density 1.1607
Quantity 250 g
Formula Weight 75.07
Percent Purity ≥99.5%
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM31L033 Certificate TypeCertificate of Analysis Date03/02/2025 Catalog Number036435.30,036435.A1
Lot NumberY25K020 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number036435.30,036435.A1
Lot NumberX21K007 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number036435.30,036435.A1
Lot NumberW26K021 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number036435.30,036435.A1
Lot NumberU28K024 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number036435.30,036435.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Glycine, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.