missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Glycine, 99%, ACS reagent Product Code.: 12317153

Thermo Scientific Chemicals Glycine, 99%, ACS reagent

Product Code. 12317153
50 g, Plastic Bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 12317153

Brand: Thermo Scientific Chemicals 385900500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula H2NCH2CO2H
Density 1.5950g/mL
Quantity 50 g
Formula Weight 75.07
Grade ACS Reagent
Chemical Name or Material Glycine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466388 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number385900500,385902500
Lot NumberA0457451 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number385900500,385902500
Lot NumberA0454283 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number385900500,385902500
Lot NumberA0451468 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number385900500,385902500
Lot NumberA0397320 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number385900500,385902500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Glycine, 99%, ACS reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.