missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 99+%, for analysis

Glycine, CAS # 56-40-6, is a purified formulation of the amino acid, glycine, (aminoacetic acid) that is used in the synthesis of several compounds including creatinine, purines and porphyrins.

41.00 € - 1390.92 €

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10733031
View Documents
Thermo Scientific Chemicals
220911000
100 g Plastic Bottle
41.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
10072930
View Documents
Thermo Scientific Chemicals
220912500
250 g Plastic Bottle
43.62 €
250g
Please to purchase this item. Need a web account? Register with us today!
10358210
View Documents
Thermo Scientific Chemicals
220910010
1 kg Plastic Bottle
103.52 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10773644
View Documents
Thermo Scientific Chemicals
220910050
5 kg Plastic Drum
364.82 €
5kg
Please to purchase this item. Need a web account? Register with us today!
11945341
View Documents
Thermo Scientific Chemicals
220910250
25 kg Fiber Drum
1390.92 €
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Glycine is an essential acid that can be used for synthesis of creatine, purines and porphyrins
  • Soluble in water; has a pH of ∼=5.5-6.5 in solution
  • Glycine is an inhibitory neurotransmitter that regulates the activity of NMDA receptors and affects neural activity

Applications

  • Glycine may also be used to maintain osmotic pressure, or as a component of other molecular biology buffers that are ion-dependent
  • Glycine is used in molecular biology as a buffer component for gel electrophoresis; the ability of glycine to adopt a negative charge at higher pH makes it ideal for separating proteins in the stacking layer
  • Intended for biochemistry applications
Specifications

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O

Specifications

200ppm max.
1ppm max.
40ppm max.
10ppm max.
10ppm max.
0.2% (105°C, 2 hrs) max.
H2NCH2CO2H
Meets the requirements
0.1% max.
DHMQDGOQFOQNFH-UHFFFAOYSA-N
NCC(O)=O
75.07
CHEBI:15428
≥99.0% (on dried substance) (total base)
Crystalline Powder
56-40-6
White
Clear solution
Authentic
99+%
C2H5NO2
MFCD00008131
50ppm max.
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
1.5950g/mL
2-aminoacetic acid
750
75.07
Analytical
Glycine
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.