missing translation for 'onlineSavingsMsg'
Learn More

Glycine, ≥99%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Product Code. 15815158
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.067
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES C(C(=O)O)N

Specifications

Melting Point ∼245°C (decomposition)
Packaging Plastic Bottle
Linear Formula H2NCH2CO2H
Beilstein 635782
Density 1.595
Quantity 500 g
Formula Weight 75.07
Percent Purity ≥99%
Specific Gravity 1.595
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.