missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glycine anhydride, 98%
Click to view available options
Quantity:
25 g
100 g
Description
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.
Solubility
Soluble in water (142mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.
Solubility
Soluble in water (142mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | ∼300°C (decomposition) |
Beilstein | 112112 |
Merck Index | 14,7466 |
Solubility Information | Soluble in water (142mg/L). |
Quantity | 25 g |
Formula Weight | 114.1 |
Percent Purity | 99% |
Chemical Name or Material | Glycine anhydride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Glycine anhydride, 98% >
Spot an opportunity for improvement?Share a Content Correction