missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Certified AR for Analysis, Fisher Chemical™

Product Code. 10589760
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

C2H5NO2, CAS Number-56-40-6, glycolixir, padil, aciport, hampshire glycine, aminoethanoic acid, glycine, glycocoll, glycosthene, glicoamin, aminoacetic acid, 1kg, CHEBI:15428, White, 75.07g/mol, DHMQDGOQFOQNFH-UHFFFAOYSA-N, 2-aminoacetic acid, 182 deg.C, 8131, 75.067, Plastic powder jar, 5.9 to 6.4

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 182°C
pH 5.9 to 6.4
Color White
Packaging Plastic Powder Jar
Formula Weight 75.07g/mol
Quantity 250 g
Physical Form Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'