missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
1368.09 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
25KG POLY BOTTLE
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 56-40-6 |
pH | 5.9 to 6.4 |
Chloride | Max. 50 ppm |
Heavy Metals (as Pb) | Max. 10 ppm |
Molecular Formula | C2H5NO2 |
Loss on Drying | Max. 0.2 % |
Packaging | Plastic Bottle |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
Residual Solvents | Complying |
SMILES | NCC(O)=O |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'