missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Honeywell Fluka™

Product Code. 16635823
Click to view available options
Quantity:
250 g
1 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Ammonium Max. 0.02 %
pH 5.9 to 6.4
Chloride Max. 30 ppm
Heavy Metals (as Pb) Max. 10 ppm
Iron (Fe) Max. 0.0005 %
Loss on Drying Max. 0.5 %
Packaging Fiberboard Box
Sulfate Max. 0.002 %
Percent Purity 99.7 to 101.0 %
Sulfated Ash Max. 0.05 %
Grade Reagent Pharmacopeia Organics
Water Max. 0.1 %
Quantity 50 kg
Physical Form Solid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.