missing translation for 'onlineSavingsMsg'
Learn More

Glycine, PlusOne™, Cytiva

Product Code. 11545005
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Use as a component of electrophoresis buffers. Glycine, PlusOne™, Cytiva is often a component of electrophoresis buffers, especially in the Laemmli discontinuous buffer system for SDS-PAGE.

  • Used in IEF and PAGE
  • Soluble in cold water and hot water, Slightly soluble in acetone
  • Insoluble in diethyl ether
  • Solid
  • Used as a component of buffers
TRUSTED_SUSTAINABILITY

Specifications

CAS 56-40-6, 7732-18-5
Assay Percent Range ≥99.7%
Molecular Formula C2H5NO2
MDL Number MFCD00008131
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES NCC(O)=O
IUPAC Name 2-aminoacetic acid
Molecular Weight (g/mol) 75.07
PubChem CID 750
ChEBI CHEBI:15428
Formula Weight 75.07
Quantity 500 g
Chemical Name or Material Glycine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'