missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glycine, sodium salt hydrate, 99%
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 197.0°C to 201.0°C |
pH | 11.1 to 12.0 (10% aq. soln.) |
Color | White to Yellow |
Assay Percent Range | 99% |
Infrared Spectrum | Authentic |
Linear Formula | H2NCH2CO2Na |
Solubility Information | Solubility in water: >500g/L (20°C). Other solubilities: solubility in methanol: 60g/L (20°C) |
IUPAC Name | sodium 2-aminoacetate |
Formula Weight | 97.05 |
Transmittance | 94.0 % min. (c=5, H2O, 400nm) 1 cm cell |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'